Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HDX
Entrez Gene ID
139324
Gene Name
highly divergent homeobox
Gene Aliases
CXorf43, D030011N01Rik
Chromosome Location
Chr.X: 84317874 - 84502578 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177478.1 NP_001170949.1 3 - 4 1358 125
NM_001177479.1 NP_001170950.1 4 - 5 1505 125
NM_144657.4 NP_653258.2 3 - 4 1396 125
XM_006724619.1 XP_006724682.1 2 - 3 1249 125
XM_011530863.2 XP_011529165.1 4 - 5 1578 125
XM_011530864.1 XP_011529166.1 4 - 5 1485 125
XM_017029274.1 XP_016884763.1 3 - 4 1376 125
GenBank mRNA AK055240.1 - 3 - 4 1332 125
AK290050.1 - 3 - 4 1370 125
BC101720.1 - 3 - 4 1308 125
BC101722.1 - 3 - 4 1308 125
BC143522.1 - 4 - 5 1417 125
BX538112.1 - 3 - 4 1368 125
BX649166.1 - 3 - 4 1371 125

View Full Product Details