Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS39
Entrez Gene ID
23339
Gene Name
VPS39, HOPS complex subunit
Gene Aliases
TLP, VAM6, hVam6p
Chromosome Location
Chr.15: 42158701 - 42208331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301138.1 NP_001288067.1 5 - 6 450 106
NM_015289.3 NP_056104.2 4 - 5 417 106
XM_011521403.2 XP_011519705.1 5 - 6 419 106
XM_011521404.2 XP_011519706.1 4 - 5 422 106
XM_017022036.1 XP_016877525.1 5 - 6 418 106
XM_017022037.1 XP_016877526.1 4 - 5 422 106
XM_017022038.1 XP_016877527.1 4 - 5 605 106
XM_017022039.1 XP_016877528.1 4 - 5 390 106
GenBank mRNA AB018313.2 - 4 - 5 368 106
AF280814.1 - 5 - 6 277 106
AF281052.1 - 4 - 5 387 106
BC068559.1 - 4 - 5 386 106
BX537404.1 - 4 - 5 396 106

View Full Product Details