Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS39
Entrez Gene ID
23339
Gene Name
VPS39, HOPS complex subunit
Gene Aliases
TLP, VAM6, hVam6p
Chromosome Location
Chr.15: 42158701 - 42208331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301138.1 NP_001288067.1 17 - 18 1885 67
NM_015289.3 NP_056104.2 16 - 17 1852 67
XM_011521403.2 XP_011519705.1 17 - 18 1854 67
XM_011521404.2 XP_011519706.1 16 - 17 1857 67
XM_017022036.1 XP_016877525.1 17 - 18 1853 67
XM_017022037.1 XP_016877526.1 16 - 17 1857 67
XM_017022038.1 XP_016877527.1 16 - 17 2040 67
XM_017022039.1 XP_016877528.1 16 - 17 1851 67
GenBank mRNA AB018313.2 - 16 - 17 1803 67
AF280814.1 - 17 - 18 1712 67
AF281052.1 - 16 - 17 1822 67
AK025334.1 - 1 - 2 52 67
AK128403.1 - 1 - 2 60 67
BC015817.2 - 12 - 13 1412 67
BC068559.1 - 16 - 17 1821 67
BX537404.1 - 16 - 17 1831 67

View Full Product Details