The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301138.1 | NP_001288067.1 | 22 - 23 | 2383 | 84 |
NM_015289.3 | NP_056104.2 | 21 - 22 | 2350 | 84 | |
XM_011521403.2 | XP_011519705.1 | 22 - 23 | 2352 | 84 | |
XM_011521404.2 | XP_011519706.1 | 21 - 22 | 2355 | 84 | |
XM_017022036.1 | XP_016877525.1 | 22 - 23 | 2351 | 84 | |
XM_017022037.1 | XP_016877526.1 | 21 - 22 | 2355 | 84 | |
XM_017022038.1 | XP_016877527.1 | 21 - 22 | 2538 | 84 | |
XM_017022039.1 | XP_016877528.1 | 21 - 22 | 2349 | 84 | |
GenBank mRNA | AB018313.2 | - | 21 - 22 | 2301 | 84 |
AF280814.1 | - | 22 - 23 | 2210 | 84 | |
AF281052.1 | - | 21 - 22 | 2320 | 84 | |
AK025334.1 | - | 6 - 7 | 550 | 84 | |
BC015817.2 | - | 17 - 18 | 1910 | 84 | |
BC068559.1 | - | 21 - 22 | 2319 | 84 | |
BX537404.1 | - | 21 - 22 | 2329 | 84 |