Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS39
Entrez Gene ID
23339
Gene Name
VPS39, HOPS complex subunit
Gene Aliases
TLP, VAM6, hVam6p
Chromosome Location
Chr.15: 42158701 - 42208331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301138.1 NP_001288067.1 10 - 11 1042 82
NM_015289.3 NP_056104.2 9 - 10 1009 82
XM_011521403.2 XP_011519705.1 10 - 11 1011 82
XM_011521404.2 XP_011519706.1 9 - 10 1014 82
XM_017022036.1 XP_016877525.1 10 - 11 1010 82
XM_017022037.1 XP_016877526.1 9 - 10 1014 82
XM_017022038.1 XP_016877527.1 9 - 10 1197 82
XM_017022039.1 XP_016877528.1 9 - 10 982 82
GenBank mRNA AB018313.2 - 9 - 10 960 82
AF280814.1 - 10 - 11 869 82
AF281052.1 - 9 - 10 979 82
BC015817.2 - 5 - 6 569 82
BC068559.1 - 9 - 10 978 82
BX537404.1 - 9 - 10 988 82

View Full Product Details