The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013386.4 | NP_037518.3 | 2 - 3 | 526 | 137 |
NM_213651.2 | NP_998816.1 | 2 - 3 | 437 | 137 | |
GenBank mRNA | AF123303.1 | - | 2 - 3 | 285 | 137 |
AJ619961.1 | - | 2 - 3 | 369 | 137 | |
AJ619987.1 | - | 2 - 3 | 315 | 137 | |
AK292567.1 | - | 2 - 3 | 437 | 137 | |
AK304165.1 | - | 2 - 3 | 520 | 137 | |
AK316304.1 | - | 2 - 3 | 480 | 137 | |
AK316506.1 | - | 2 - 3 | 348 | 137 | |
BC014519.1 | - | 2 - 3 | 382 | 137 | |
BQ685792.1 | - | 2 - 3 | 444 | 137 |