The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013386.4 | NP_037518.3 | 8 - 9 | 1311 | 145 |
NM_213651.2 | NP_998816.1 | 8 - 9 | 1222 | 145 | |
XM_017001104.1 | XP_016856593.1 | 4 - 5 | 567 | 145 | |
GenBank mRNA | AF123303.1 | - | 8 - 9 | 1070 | 145 |
AJ619961.1 | - | 8 - 9 | 1154 | 145 | |
AJ619987.1 | - | 8 - 9 | 1100 | 145 | |
AK292567.1 | - | 8 - 9 | 1222 | 145 | |
AK304165.1 | - | 8 - 9 | 1305 | 145 | |
AK316304.1 | - | 8 - 9 | 1265 | 145 | |
AK316506.1 | - | 8 - 9 | 1133 | 145 | |
BC014519.1 | - | 8 - 9 | 1167 | 145 | |
BC068561.1 | - | 8 - 9 | 1200 | 145 |