The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199417.1 | NP_001186346.1 | 15 - 16 | 2763 | 70 |
XM_006721991.1 | XP_006722054.1 | 15 - 16 | 2804 | 70 | |
XM_006721992.3 | XP_006722055.1 | 15 - 16 | 2542 | 70 | |
XM_006721993.3 | XP_006722056.1 | 15 - 16 | 2790 | 70 | |
XM_006721994.3 | XP_006722057.1 | 15 - 16 | 2789 | 70 | |
XM_011525073.1 | XP_011523375.1 | 15 - 16 | 2723 | 70 | |
XM_011525074.2 | XP_011523376.1 | 15 - 16 | 2822 | 70 | |
XM_011525075.2 | XP_011523377.1 | 10 - 11 | 2364 | 70 | |
XM_011525076.2 | XP_011523378.1 | 15 - 16 | 2789 | 70 | |
XM_011525077.2 | XP_011523379.1 | 15 - 16 | 2789 | 70 | |
XM_017024902.1 | XP_016880391.1 | 15 - 16 | 2789 | 70 | |
GenBank mRNA | AB040934.1 | - | 9 - 10 | 1526 | 70 |
BC039190.1 | - | 8 - 9 | 1036 | 70 | |
DN831435.1 | - | 2 - 3 | 239 | 70 |