The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004599.3 | NP_004590.2 | 13 - 14 | 2685 | 108 |
NR_103834.1 | 14 - 15 | 2785 | 108 | ||
XM_006724310.2 | XP_006724373.1 | 13 - 14 | 2929 | 108 | |
XM_011530347.1 | XP_011528649.1 | 11 - 12 | 2139 | 108 | |
XM_017028921.1 | XP_016884410.1 | 13 - 14 | 2514 | 108 | |
GenBank mRNA | AK299914.1 | - | 13 - 14 | 2514 | 108 |
AL834221.1 | - | 5 - 6 | 1044 | 108 | |
BC051385.1 | - | 14 - 15 | 2691 | 108 | |
BC051799.1 | - | 13 - 14 | 2651 | 108 | |
BC056158.1 | - | 13 - 14 | 2631 | 108 | |
CT841522.1 | - | 13 - 14 | 2575 | 108 | |
U02031.1 | - | 13 - 14 | 2610 | 108 |