The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271539.1 | NP_001258468.1 | 13 - 14 | 1246 | 100 |
NR_073206.1 | 14 - 15 | 1367 | 100 | ||
NR_073207.1 | 13 - 14 | 1414 | 100 | ||
XM_006723646.2 | XP_006723709.1 | 13 - 14 | 1248 | 100 | |
XM_006723647.2 | XP_006723710.1 | 13 - 14 | 1248 | 100 | |
XM_006723648.2 | XP_006723711.1 | 12 - 13 | 1191 | 100 | |
XM_006723650.2 | XP_006723713.1 | 13 - 14 | 1248 | 100 | |
XM_006723651.2 | XP_006723714.1 | 13 - 14 | 1248 | 100 | |
XM_006723652.2 | XP_006723715.1 | 13 - 14 | 1248 | 100 | |
XM_006723654.3 | XP_006723717.1 | 4 - 5 | 263 | 100 | |
XM_011529374.1 | XP_011527676.1 | 12 - 13 | 1188 | 100 | |
XM_011529375.1 | XP_011527677.1 | 13 - 14 | 1137 | 100 | |
XM_011529376.1 | XP_011527678.1 | 12 - 13 | 1080 | 100 | |
XM_011529377.1 | XP_011527679.1 | 14 - 15 | 1457 | 100 | |
XM_011529378.1 | XP_011527680.1 | 11 - 12 | 966 | 100 | |
XM_011529379.1 | XP_011527681.1 | 11 - 12 | 999 | 100 | |
XM_017028084.1 | XP_016883573.1 | 2 - 3 | 185 | 100 | |
GenBank mRNA | AK302170.1 | - | 13 - 14 | 1243 | 100 |
DA857535.1 | - | 1 - 2 | 38 | 100 |