The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271539.1 | NP_001258468.1 | 3 - 4 | 371 | 142 |
NR_073206.1 | 3 - 4 | 371 | 142 | ||
NR_073207.1 | 3 - 4 | 539 | 142 | ||
XM_006723646.2 | XP_006723709.1 | 3 - 4 | 373 | 142 | |
XM_006723647.2 | XP_006723710.1 | 3 - 4 | 373 | 142 | |
XM_006723648.2 | XP_006723711.1 | 3 - 4 | 373 | 142 | |
XM_006723650.2 | XP_006723713.1 | 3 - 4 | 373 | 142 | |
XM_006723651.2 | XP_006723714.1 | 3 - 4 | 373 | 142 | |
XM_006723652.2 | XP_006723715.1 | 3 - 4 | 373 | 142 | |
XM_006723655.2 | XP_006723718.1 | 3 - 4 | 373 | 142 | |
XM_006723656.2 | XP_006723719.1 | 3 - 4 | 373 | 142 | |
XM_011529374.1 | XP_011527676.1 | 3 - 4 | 373 | 142 | |
XM_011529375.1 | XP_011527677.1 | 3 - 4 | 373 | 142 | |
XM_011529376.1 | XP_011527678.1 | 3 - 4 | 373 | 142 | |
GenBank mRNA | AK302170.1 | - | 3 - 4 | 368 | 142 |
DC396776.1 | - | 3 - 4 | 371 | 142 | |
DC398207.1 | - | 3 - 4 | 368 | 142 |