The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001819.2 | NP_001810.2 | 3 - 4 | 498 | 112 |
GenBank mRNA | AB209712.1 | - | 3 - 4 | 295 | 112 |
AK225125.1 | - | 3 - 4 | 316 | 112 | |
AK289386.1 | - | 3 - 4 | 318 | 112 | |
AK312237.1 | - | 3 - 4 | 318 | 112 | |
BC000375.2 | - | 3 - 4 | 296 | 112 | |
CR456726.1 | - | 3 - 4 | 192 | 112 | |
Y00064.1 | - | 3 - 4 | 304 | 112 |