Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TDRKH
Entrez Gene ID
11022
Gene Name
tudor and KH domain containing
Gene Aliases
TDRD2
Chromosome Location
Chr.1: 151769354 - 151790576 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083963.1 NP_001077432.1 2 - 3 282 105
NM_001083964.1 NP_001077433.1 2 - 3 307 105
NM_001083965.1 NP_001077434.1 2 - 3 307 105
NM_006862.3 NP_006853.2 2 - 3 307 105
XM_005244856.4 XP_005244913.1 2 - 3 173 105
XM_017000122.1 XP_016855611.1 2 - 3 349 105
XM_017000123.1 XP_016855612.1 2 - 3 349 105
XM_017000124.1 XP_016855613.1 2 - 3 248 105
XM_017000125.1 XP_016855614.1 2 - 3 277 105
XM_017000126.1 XP_016855615.1 2 - 3 248 105
XM_017000127.1 XP_016855616.1 2 - 3 277 105
XM_017000128.1 XP_016855617.1 2 - 3 302 105
XM_017000129.1 XP_016855618.1 2 - 3 267 105
XM_017000130.1 XP_016855619.1 2 - 3 241 105
GenBank mRNA AF119121.2 - 2 - 3 295 105
AF227192.1 - 2 - 3 295 105
AK056402.1 - 2 - 3 242 105
AK226092.1 - 2 - 3 220 105
AK295948.1 - 2 - 3 178 105
AK303063.1 - 2 - 3 203 105
AK316279.1 - 2 - 3 258 105
BC032690.1 - 2 - 3 214 105
DA796638.1 - 2 - 3 307 105

View Full Product Details