Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TDRKH
Entrez Gene ID
11022
Gene Name
tudor and KH domain containing
Gene Aliases
TDRD2
Chromosome Location
Chr.1: 151769354 - 151790576 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083963.1 NP_001077432.1 5 - 6 714 108
NM_001083964.1 NP_001077433.1 5 - 6 604 108
NM_001083965.1 NP_001077434.1 5 - 6 739 108
NM_006862.3 NP_006853.2 5 - 6 739 108
XM_005244856.4 XP_005244913.1 5 - 6 505 108
XM_017000122.1 XP_016855611.1 5 - 6 781 108
XM_017000123.1 XP_016855612.1 5 - 6 781 108
XM_017000124.1 XP_016855613.1 5 - 6 680 108
XM_017000125.1 XP_016855614.1 5 - 6 709 108
XM_017000126.1 XP_016855615.1 5 - 6 680 108
XM_017000127.1 XP_016855616.1 5 - 6 709 108
XM_017000128.1 XP_016855617.1 5 - 6 634 108
XM_017000129.1 XP_016855618.1 5 - 6 599 108
XM_017000130.1 XP_016855619.1 5 - 6 573 108
GenBank mRNA AF119121.2 - 5 - 6 727 108
AF227192.1 - 5 - 6 727 108
AK226092.1 - 5 - 6 652 108
AK295948.1 - 5 - 6 510 108
AK303063.1 - 5 - 6 635 108
AK316279.1 - 5 - 6 590 108
BC032690.1 - 5 - 6 646 108

View Full Product Details