Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TDRKH
Entrez Gene ID
11022
Gene Name
tudor and KH domain containing
Gene Aliases
TDRD2
Chromosome Location
Chr.1: 151769354 - 151790576 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083963.1 NP_001077432.1 7 - 8 1201 85
NM_001083964.1 NP_001077433.1 7 - 8 1091 85
NM_001083965.1 NP_001077434.1 7 - 8 1226 85
NM_006862.3 NP_006853.2 7 - 8 1226 85
XM_005244856.4 XP_005244913.1 7 - 8 992 85
XM_017000122.1 XP_016855611.1 7 - 8 1268 85
XM_017000123.1 XP_016855612.1 7 - 8 1268 85
XM_017000124.1 XP_016855613.1 7 - 8 1167 85
XM_017000125.1 XP_016855614.1 7 - 8 1196 85
XM_017000126.1 XP_016855615.1 7 - 8 1167 85
XM_017000127.1 XP_016855616.1 7 - 8 1196 85
XM_017000128.1 XP_016855617.1 7 - 8 1121 85
XM_017000129.1 XP_016855618.1 7 - 8 1086 85
XM_017000130.1 XP_016855619.1 7 - 8 1060 85
GenBank mRNA AF119121.2 - 7 - 8 1214 85
AF227192.1 - 7 - 8 1214 85
AK056402.1 - 7 - 8 1149 85
AK225160.1 - 7 - 8 1080 85
AK226092.1 - 7 - 8 1139 85
AK295948.1 - 7 - 8 997 85
AK303063.1 - 7 - 8 1122 85
BC032690.1 - 7 - 8 1133 85

View Full Product Details