The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014763.1 | NP_001014763.1 | 3 - 4 | 915 | 105 |
NM_001985.2 | NP_001976.1 | 4 - 5 | 530 | 105 | |
GenBank mRNA | AF370381.1 | - | 3 - 4 | 897 | 105 |
AK055285.1 | - | 3 - 4 | 917 | 105 | |
AK291881.1 | - | 4 - 5 | 457 | 105 | |
AL833205.1 | - | 3 - 4 | 3191 | 105 | |
BC093961.1 | - | 4 - 5 | 461 | 105 | |
BC093963.1 | - | 4 - 5 | 460 | 105 | |
BG281641.1 | - | 3 - 4 | 348 | 105 | |
BG399783.1 | - | 4 - 5 | 531 | 105 | |
CR456827.1 | - | 4 - 5 | 438 | 105 | |
X71129.1 | - | 4 - 5 | 465 | 105 |