The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020745.3 | NP_065796.1 | 15 - 16 | 2149 | 116 |
XM_005249245.3 | XP_005249302.1 | 13 - 14 | 1890 | 116 | |
XM_011514764.2 | XP_011513066.1 | 15 - 16 | 2181 | 116 | |
XM_017011112.1 | XP_016866601.1 | 13 - 14 | 1731 | 116 | |
GenBank mRNA | AB033096.1 | - | 15 - 16 | 2149 | 116 |
BC013593.1 | - | 6 - 7 | 844 | 116 | |
BC033169.1 | - | 12 - 13 | 1546 | 116 | |
BC131728.1 | - | 15 - 16 | 2149 | 116 |