The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020745.3 | NP_065796.1 | 17 - 18 | 2369 | 80 |
XM_005249245.3 | XP_005249302.1 | 15 - 16 | 2110 | 80 | |
XM_011514764.2 | XP_011513066.1 | 17 - 18 | 2401 | 80 | |
XM_017011112.1 | XP_016866601.1 | 15 - 16 | 1951 | 80 | |
GenBank mRNA | AB033096.1 | - | 17 - 18 | 2369 | 80 |
BC013593.1 | - | 8 - 9 | 1064 | 80 | |
BC033169.1 | - | 14 - 15 | 1766 | 80 | |
BC131728.1 | - | 17 - 18 | 2369 | 80 | |
CN343332.1 | - | 2 - 3 | 171 | 80 |