The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032410.3 | NP_115786.1 | 12 - 13 | 1478 | 91 |
GenBank mRNA | AK090540.1 | - | 2 - 3 | 202 | 91 |
AK309224.1 | - | 12 - 13 | 1422 | 91 | |
BC048304.1 | - | 12 - 13 | 1495 | 91 | |
BC056146.1 | - | 12 - 13 | 1359 | 91 |