Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGCB
Entrez Gene ID
6443
Gene Name
sarcoglycan beta
Gene Aliases
A3b, LGMD2E, SGC
Chromosome Location
Chr.4: 52020695 - 52038319 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000232.4 NP_000223.1 2 - 3 294 79
GenBank mRNA AK289892.1 - 2 - 3 272 79
BC020709.1 - 2 - 3 255 79
BT019433.1 - 2 - 3 234 79
CN483961.1 - 2 - 3 294 79
CR456810.1 - 2 - 3 234 79
U29586.1 - 2 - 3 234 79
U31116.1 - 2 - 3 276 79

View Full Product Details