Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DCAF4
- Entrez Gene ID
-
26094
- Gene Name
- DDB1 and CUL4 associated factor 4
- Gene Aliases
-
WDR21, WDR21A
- Chromosome Location
-
Chr.14: 72926332 - 72962098 on Build GRCh38
View Full Product Details