Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LMO4
Entrez Gene ID
8543
Gene Name
LIM domain only 4
Gene Aliases
-
Chromosome Location
Chr.1: 87328468 - 87348923 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006769.3 NP_006760.1 3 - 4 1117 66
XM_005271291.3 XP_005271348.1 3 - 4 3287 66
GenBank mRNA AK292578.1 - 3 - 4 719 66
BC003600.1 - 3 - 4 480 66
BC017673.1 - 3 - 4 605 66
BC065818.1 - 3 - 4 411 66
U24576.1 - 3 - 4 1117 66

View Full Product Details