Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZACN
Entrez Gene ID
353174
Gene Name
zinc activated ion channel
Gene Aliases
L2, LGICZ, LGICZ1, ZAC, ZAC1
Chromosome Location
Chr.17: 76079182 - 76082804 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_180990.3 NP_851321.2 4 - 5 461 84
GenBank mRNA AB223030.1 - 4 - 5 375 84
AF512521.1 - 4 - 5 415 84
AK122638.1 - 4 - 5 461 84
BC110597.1 - 4 - 5 415 84
DB119720.1 - 4 - 5 461 84

View Full Product Details