The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005246024.4 | XP_005246081.1 | 1 - 2 | 142 | 80 |
XM_017002670.1 | XP_016858159.1 | 1 - 2 | 142 | 80 | |
GenBank mRNA | AF022726.2 | - | 1 - 2 | 105 | 80 |