The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015960.2 | NP_057044.2 | 2 - 3 | 285 | 108 |
GenBank mRNA | AF132966.1 | - | 2 - 3 | 239 | 108 |
AK001142.1 | - | 2 - 3 | 205 | 108 | |
AK302507.1 | - | 2 - 3 | 221 | 108 | |
AK314687.1 | - | 2 - 3 | 221 | 108 | |
BC015059.1 | - | 2 - 3 | 182 | 108 | |
BC021105.1 | - | 2 - 3 | 164 | 108 | |
BC028948.1 | - | 2 - 3 | 253 | 108 | |
CD107559.1 | - | - | 311 | 108 |