The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024667.2 | NP_078943.1 | 2 - 3 | 384 | 73 |
XM_005253622.3 | XP_005253679.1 | 3 - 4 | 3099 | 73 | |
XM_006719602.3 | XP_006719665.1 | 3 - 4 | 420 | 73 | |
XM_011538743.2 | XP_011537045.1 | 3 - 4 | 525 | 73 | |
XM_011538745.2 | XP_011537047.1 | 2 - 3 | 2915 | 73 | |
GenBank mRNA | AK022812.1 | - | 2 - 3 | 384 | 73 |
BC005882.2 | - | 2 - 3 | 322 | 73 |