The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032410.3 | NP_115786.1 | 19 - 20 | 2079 | 99 |
GenBank mRNA | AK090540.1 | - | 9 - 10 | 803 | 99 |
BC056146.1 | - | 19 - 20 | 1960 | 99 |