Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNIP1
Entrez Gene ID
96459
Gene Name
folliculin interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.5: 131641714 - 131797063 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008738.2 NP_001008738.2 9 - 10 1173 85
NM_133372.2 NP_588613.2 10 - 11 1257 85
GenBank mRNA AB075841.1 - 4 - 5 446 85
AK292473.1 - 10 - 11 1211 85
BC001956.1 - 10 - 11 1210 85
CR749426.1 - 4 - 5 496 85
CR936723.1 - 9 - 10 1010 85
DQ145719.1 - 10 - 11 1162 85

View Full Product Details