Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DBN1
Entrez Gene ID
1627
Gene Name
drebrin 1
Gene Aliases
D0S117E
Chromosome Location
Chr.5: 177456612 - 177480368 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004395.3 NP_004386.2 12 - 13 1947 66
NM_080881.2 NP_543157.1 13 - 14 2063 66
XM_005265827.3 XP_005265884.1 13 - 14 2032 66
XM_006714826.2 XP_006714889.1 13 - 14 1922 66
XM_011534446.2 XP_011532748.1 14 - 15 2204 66
XM_011534447.2 XP_011532749.1 13 - 14 2701 66
XM_017009139.1 XP_016864628.1 13 - 14 2203 66
XM_017009140.1 XP_016864629.1 12 - 13 2056 66
GenBank mRNA AK094125.1 - 11 - 12 1993 66
AK225523.1 - 12 - 13 1888 66
AK295295.1 - 5 - 6 1162 66
AK314645.1 - 12 - 13 1888 66
AL110225.1 - 13 - 14 2392 66
BC000283.1 - 12 - 13 1871 66
BC007281.1 - 12 - 13 1871 66
BC007567.1 - 12 - 13 1871 66
D17530.1 - 12 - 13 1872 66
U00802.1 - 12 - 13 1847 66

View Full Product Details