Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DBN1
Entrez Gene ID
1627
Gene Name
drebrin 1
Gene Aliases
D0S117E
Chromosome Location
Chr.5: 177456612 - 177480368 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080881.2 NP_543157.1 2 - 3 379 66
XM_011534446.2 XP_011532748.1 2 - 3 382 66
XM_017009139.1 XP_016864628.1 1 - 2 381 66
XM_017009140.1 XP_016864629.1 1 - 2 372 66
GenBank mRNA AL110225.1 - 2 - 3 708 66

View Full Product Details