The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005265827.3 | XP_005265884.1 | 10 - 11 | 1076 | 65 |
XM_006714826.2 | XP_006714889.1 | 10 - 11 | 966 | 65 | |
XM_011534446.2 | XP_011532748.1 | 11 - 12 | 1248 | 65 | |
XM_011534447.2 | XP_011532749.1 | 10 - 11 | 1745 | 65 | |
XM_017009139.1 | XP_016864628.1 | 10 - 11 | 1247 | 65 |