The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040454.1 | NP_001035544.1 | 8 - 9 | 1320 | 113 |
NM_001281732.1 | NP_001268661.1 | 8 - 9 | 1124 | 113 | |
NM_001281733.1 | NP_001268662.1 | 7 - 8 | 911 | 113 | |
NM_022911.2 | NP_075062.2 | 9 - 10 | 1232 | 113 | |
NM_134263.2 | NP_599025.2 | 9 - 10 | 1232 | 113 | |
NM_134426.2 | NP_602298.2 | 9 - 10 | 1232 | 113 | |
GenBank mRNA | AB033288.1 | - | 9 - 10 | 1195 | 113 |
AB102713.1 | - | 9 - 10 | 1277 | 113 | |
AB208936.1 | - | 3 - 4 | 3081 | 113 | |
AF279265.1 | - | 8 - 9 | 1320 | 113 | |
AF288410.1 | - | 9 - 10 | 1180 | 113 | |
AF416721.1 | - | 9 - 10 | 1180 | 113 | |
AK056237.1 | - | 7 - 8 | 911 | 113 | |
AK297695.1 | - | 8 - 9 | 1072 | 113 | |
BC017697.1 | - | 9 - 10 | 1181 | 113 |