The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040454.1 | NP_001035544.1 | 15 - 16 | 1989 | 80 |
NM_001281732.1 | NP_001268661.1 | 15 - 16 | 1793 | 80 | |
NM_001281733.1 | NP_001268662.1 | 14 - 15 | 1580 | 80 | |
NM_022911.2 | NP_075062.2 | 16 - 17 | 1901 | 80 | |
NM_134263.2 | NP_599025.2 | 16 - 17 | 1901 | 80 | |
GenBank mRNA | AF279265.1 | - | 15 - 16 | 1989 | 80 |
AF288410.1 | - | 16 - 17 | 1849 | 80 | |
AF416721.1 | - | 16 - 17 | 1849 | 80 | |
AK056237.1 | - | 14 - 15 | 1580 | 80 | |
AK297695.1 | - | 15 - 16 | 1741 | 80 | |
BC017697.1 | - | 16 - 17 | 1850 | 80 |