Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOP10
Entrez Gene ID
55505
Gene Name
NOP10 ribonucleoprotein
Gene Aliases
DKCB1, NOLA3, NOP10P
Chromosome Location
Chr.15: 34341716 - 34343161 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018648.3 NP_061118.1 2 - 2 382 118
GenBank mRNA AB043104.1 - 2 - 2 392 118
BC008886.2 - 2 - 2 327 118
BC063023.1 - 2 - 2 353 118
BM554889.1 - 2 - 2 323 118
BQ218536.1 - 2 - 2 398 118

View Full Product Details