Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPST2
Entrez Gene ID
8459
Gene Name
tyrosylprotein sulfotransferase 2
Gene Aliases
TANGO13B
Chromosome Location
Chr.22: 26524082 - 26590125 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008566.1 NP_001008566.1 5 - 6 1430 59
NM_003595.3 NP_003586.3 5 - 6 1322 59
XM_006724338.3 XP_006724401.1 6 - 7 1441 59
GenBank mRNA AF049891.1 - 5 - 6 1286 59
AF061254.1 - 3 - 4 1094 59
AJ006198.1 - 3 - 4 1176 59
AK074538.1 - 5 - 6 1322 59
AK075139.1 - 5 - 6 1394 59
AK304545.1 - 7 - 8 1406 59
AL136623.2 - 5 - 6 1306 59
BC001057.2 - 5 - 6 1301 59
BC017509.1 - 5 - 6 1286 59
CR456597.1 - 3 - 4 1184 59
CR533528.1 - 3 - 4 1089 59

View Full Product Details