The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013438.2 | NP_001013456.1 | 12 - 13 | 1370 | 68 |
NM_001013439.2 | NP_001013457.1 | 13 - 14 | 1714 | 68 | |
NM_005087.3 | NP_005078.2 | 12 - 13 | 1370 | 68 | |
XM_005247815.3 | XP_005247872.2 | 12 - 13 | 1145 | 68 | |
XM_017007299.1 | XP_016862788.1 | 13 - 14 | 1488 | 68 | |
XM_017007301.1 | XP_016862790.1 | 13 - 14 | 1488 | 68 | |
GenBank mRNA | AK225334.1 | - | 12 - 13 | 1146 | 68 |
AK292633.1 | - | 12 - 13 | 1146 | 68 | |
AK294603.1 | - | 11 - 12 | 1092 | 68 | |
AK297254.1 | - | 10 - 11 | 998 | 68 | |
AK302197.1 | - | 12 - 13 | 1149 | 68 | |
AY341428.1 | - | 12 - 13 | 1421 | 68 | |
BC028983.1 | - | 12 - 13 | 1140 | 68 | |
KR259877.1 | - | 12 - 13 | 1130 | 68 | |
KR259880.1 | - | 13 - 14 | 1442 | 68 | |
U25165.1 | - | 12 - 13 | 1142 | 68 |