Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL11A1
Entrez Gene ID
1301
Gene Name
collagen type XI alpha 1 chain
Gene Aliases
CO11A1, COLL6, STL2
Chromosome Location
Chr.1: 102876467 - 103108580 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190709.1 NP_001177638.1 33 - 34 2907 140
NM_001854.3 NP_001845.3 34 - 35 3024 140
NM_080629.2 NP_542196.2 34 - 35 3060 140
NM_080630.3 NP_542197.3 32 - 33 2676 140
NR_134980.1 35 - 36 3040 140
XM_017000334.1 XP_016855823.1 35 - 36 3260 140
XM_017000335.1 XP_016855824.1 35 - 36 3254 140
XM_017000336.1 XP_016855825.1 35 - 36 3261 140
XM_017000337.1 XP_016855826.1 28 - 29 1738 140
GenBank mRNA AB208844.1 - 31 - 32 2040 140
BC117697.1 - 32 - 33 2659 140
J04177.1 - 34 - 35 2867 140

View Full Product Details