Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL11A1
Entrez Gene ID
1301
Gene Name
collagen type XI alpha 1 chain
Gene Aliases
CO11A1, COLL6, STL2
Chromosome Location
Chr.1: 102876467 - 103108580 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190709.1 NP_001177638.1 38 - 39 3231 78
NM_001854.3 NP_001845.3 39 - 40 3348 78
NM_080629.2 NP_542196.2 39 - 40 3384 78
NM_080630.3 NP_542197.3 37 - 38 3000 78
NR_134980.1 40 - 41 3364 78
XM_017000334.1 XP_016855823.1 40 - 41 3584 78
XM_017000335.1 XP_016855824.1 40 - 41 3578 78
XM_017000336.1 XP_016855825.1 40 - 41 3585 78
XM_017000337.1 XP_016855826.1 33 - 34 2062 78
GenBank mRNA AB208844.1 - 36 - 37 2364 78
BC117697.1 - 37 - 38 2983 78
J04177.1 - 39 - 40 3191 78

View Full Product Details