Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IRS2
Entrez Gene ID
8660
Gene Name
insulin receptor substrate 2
Gene Aliases
IRS-2
Chromosome Location
Chr.13: 109753837 - 109786567 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003749.2 NP_003740.2 1 - 1 3262 75
GenBank mRNA AF073310.1 - 1 - 1 3264 75

View Full Product Details