The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002519.2 | NP_002510.2 | 16 - 17 | 3168 | 136 |
GenBank mRNA | AK290020.1 | - | 16 - 17 | 3168 | 136 |
AK291687.1 | - | 16 - 17 | 3092 | 136 | |
AK301256.1 | - | 3 - 4 | 439 | 136 | |
BC040356.1 | - | 16 - 17 | 3159 | 136 | |
BC050561.2 | - | 16 - 17 | 3153 | 136 | |
BC136270.1 | - | 16 - 17 | 3089 | 136 | |
D83243.1 | - | 16 - 17 | 3132 | 136 | |
U58852.1 | - | 16 - 17 | 3066 | 136 | |
X97186.1 | - | 16 - 17 | 3100 | 136 |