Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGAP22
Entrez Gene ID
58504
Gene Name
Rho GTPase activating protein 22
Gene Aliases
RhoGAP2, RhoGap22
Chromosome Location
Chr.10: 48429831 - 48656265 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256024.1 NP_001242953.1 7 - 8 1245 123
NM_001256025.2 NP_001242954.1 7 - 8 1111 123
NM_001256026.1 NP_001242955.1 5 - 6 751 123
NM_021226.3 NP_067049.2 7 - 8 1197 123
NR_045675.1 8 - 9 1696 123
XM_005270014.3 XP_005270071.1 5 - 6 655 123
XM_011540002.1 XP_011538304.1 8 - 9 1796 123
XM_011540003.1 XP_011538305.1 8 - 9 1098 123
XM_011540004.1 XP_011538306.1 8 - 9 1748 123
XM_011540005.1 XP_011538307.1 5 - 6 799 123
XM_011540006.1 XP_011538308.1 7 - 8 1619 123
XM_011540007.2 XP_011538309.1 5 - 6 5003 123
XM_011540011.2 XP_011538313.1 4 - 5 478 123
XM_011540012.2 XP_011538314.1 5 - 6 1156 123
XM_011540013.2 XP_011538315.1 8 - 9 1796 123
XM_011540015.2 XP_011538317.1 8 - 9 1796 123
XM_017016470.1 XP_016871959.1 6 - 7 1065 123
XM_017016471.1 XP_016871960.1 5 - 6 1070 123
XM_017016472.1 XP_016871961.1 5 - 6 643 123
XM_017016473.1 XP_016871962.1 8 - 9 1748 123
XM_017016474.1 XP_016871963.1 7 - 8 1194 123
XM_017016475.1 XP_016871964.1 6 - 7 1065 123
XM_017016476.1 XP_016871965.1 7 - 8 1194 123
XM_017016477.1 XP_016871966.1 7 - 8 1142 123
GenBank mRNA AK055037.1 - 6 - 7 790 123
AK055937.1 - 8 - 9 1696 123
AK127586.1 - 5 - 6 751 123
AK293579.1 - 7 - 8 997 123
AY324801.1 - 7 - 8 1159 123
BC047096.1 - 7 - 8 1197 123
BC126444.1 - 7 - 8 1066 123
BC136319.1 - 7 - 8 1066 123
EF560749.1 - 7 - 8 1169 123

View Full Product Details