The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015718.2 | NP_056533.1 | 1 - 2 | 152 | 89 |
GenBank mRNA | AF190122.1 | - | 1 - 2 | 152 | 89 |