The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005792.2 | NP_005783.2 | 1 - 2 | 111 | |
GenBank mRNA | AK222667.1 | - | 1 - 2 | 111 | |
AK314163.1 | - | 1 - 2 | 111 | ||
AW175781.1 | - | 1 - 2 | 127 | 111 | |
BU595100.1 | - | 1 - 2 | 111 |