Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FNDC4
Entrez Gene ID
64838
Gene Name
fibronectin type III domain containing 4
Gene Aliases
FRCP1
Chromosome Location
Chr.2: 27491883 - 27495303 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022823.2 NP_073734.1 3 - 4 661 73
XM_005264499.3 XP_005264556.1 3 - 4 705 73
GenBank mRNA AK026015.1 - 3 - 4 621 73
AY358136.1 - 3 - 4 379 73
BC032725.1 - 2 - 3 292 73

View Full Product Details