Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FNDC4
Entrez Gene ID
64838
Gene Name
fibronectin type III domain containing 4
Gene Aliases
FRCP1
Chromosome Location
Chr.2: 27491883 - 27495303 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022823.2 NP_073734.1 6 - 7 1076 114
GenBank mRNA AK026015.1 - 6 - 7 1036 114
AY358136.1 - 6 - 7 794 114
BC032725.1 - 5 - 6 707 114

View Full Product Details