Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DZANK1
Entrez Gene ID
55184
Gene Name
double zinc ribbon and ankyrin repeat domains 1
Gene Aliases
ANKRD64, C20orf12, C20orf84, bA189K21.1, bA189K21.8, dJ568F9.2
Chromosome Location
Chr.20: 18383367 - 18467185 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099407.1 NP_001092877.1 20 - 21 2303 62
XM_005260742.3 XP_005260799.1 20 - 21 2355 62
XM_005260743.3 XP_005260800.1 19 - 20 2209 62
XM_006723584.2 XP_006723647.1 18 - 19 2130 62
XM_011529268.2 XP_011527570.1 20 - 21 2473 62
XM_011529271.2 XP_011527573.1 20 - 21 3086 62
XM_011529272.2 XP_011527574.1 20 - 21 2469 62
XM_011529275.2 XP_011527577.1 19 - 20 2959 62
XM_011529277.2 XP_011527579.1 12 - 13 3200 62
XM_017027909.1 XP_016883398.1 20 - 21 2456 62
XM_017027910.1 XP_016883399.1 20 - 21 2339 62
XM_017027911.1 XP_016883400.1 20 - 21 2335 62
XM_017027912.1 XP_016883401.1 20 - 21 3042 62
XM_017027913.1 XP_016883402.1 20 - 21 3027 62
XM_017027914.1 XP_016883403.1 20 - 21 2281 62
XM_017027915.1 XP_016883404.1 20 - 21 2277 62
XM_017027916.1 XP_016883405.1 18 - 19 2841 62
XM_017027917.1 XP_016883406.1 21 - 22 3136 62
XM_017027918.1 XP_016883407.1 21 - 22 2390 62
XM_017027919.1 XP_016883408.1 21 - 22 2386 62
XM_017027920.1 XP_016883409.1 19 - 20 2345 62
XM_017027921.1 XP_016883410.1 13 - 14 1507 62
GenBank mRNA AK001462.1 - 14 - 15 1490 62
AK026306.1 - 6 - 7 512 62
AK055544.1 - 19 - 20 2098 62
AK131451.1 - 22 - 23 3126 62
AK295073.1 - 20 - 21 2213 62
AK299746.1 - 20 - 21 2226 62
BC130341.1 - 19 - 20 2131 62
BC144153.1 - 19 - 20 2131 62
CR749327.1 - 21 - 22 2355 62
DQ104738.1 - 11 - 12 1224 62

View Full Product Details