The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000064.3 | NP_000055.2 | 32 - 33 | 4209 | 133 |
GenBank mRNA | BC150179.1 | - | 32 - 33 | 4148 | 133 |
BC150200.1 | - | 32 - 33 | 4148 | 133 | |
EU794602.1 | - | 32 - 33 | 4148 | 133 | |
K02765.1 | - | 32 - 33 | 4176 | 133 |