The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000064.3 | NP_000055.2 | 6 - 7 | 778 | 69 |
GenBank mRNA | AK304071.1 | - | 6 - 7 | 746 | 69 |
BC150179.1 | - | 6 - 7 | 717 | 69 | |
BC150200.1 | - | 6 - 7 | 717 | 69 | |
EU794602.1 | - | 6 - 7 | 717 | 69 | |
K02765.1 | - | 6 - 7 | 745 | 69 |