The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006143.2 | NP_006134.1 | 3 - 4 | 334 | 113 |
XM_017019216.1 | XP_016874705.1 | 3 - 4 | 806 | 113 | |
GenBank mRNA | AK074729.1 | - | 1 - 2 | 172 | 113 |
DB022290.1 | - | 3 - 4 | 355 | 113 |