The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016299.3 | NP_057383.2 | 10 - 11 | 1239 | 151 |
GenBank mRNA | AF112210.1 | - | 10 - 11 | 1058 | 151 |
AF143723.1 | - | 10 - 11 | 1048 | 151 | |
AK225265.1 | - | 10 - 11 | 1083 | 151 | |
AK225651.1 | - | 10 - 11 | 1083 | 151 | |
AK226065.1 | - | 3 - 4 | 3912 | 151 | |
AK292323.1 | - | 10 - 11 | 1047 | 151 | |
BC119690.1 | - | 10 - 11 | 1058 | 151 |